3-Isothiocyanatobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008757-1G 008757-5G
FLUO008757-1GEA 0 GBP
FLUO008757-1G FLUO008757-5G
3-Isothiocyanatobenzoic acid
3-Isothiocyanatobenzoic acid
Formula: C₈H₅NO₂S
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 164…165 °C
MDL Number: MFCD00041367
CAS Number: 2131-63-7
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR