3-Bromopropyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008751-1G
008751-5G
FLUO008751-1GEA
0
GBP
FLUO008751-1G
FLUO008751-5G
3-Bromopropyl isothiocyanate
3-Bromopropyl isothiocyanate
Formula:
BrH₂CCH₂CH₂NCS |
MDL Number:
MFCD00060421 CAS Number: 2799-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...