4-Bromo-3-methylphenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008750-5G
008750-1G
FLUO008750-5GEA
0
GBP
FLUO008750-5G
FLUO008750-1G
4-Bromo-3-methylphenyl isothiocyanate
4-Bromo-3-methylphenyl isothiocyanate
Formula:
C₈H₆BrNS |
MDL Number:
MFCD00046804 CAS Number: 71672-88-3 EINECS: 275-823-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...