4-Bromo-3-chlorophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008747-1G 008747-5G
FLUO008747-1GEA 0 GBP
FLUO008747-1G FLUO008747-5G
4-Bromo-3-chlorophenyl isothiocyanate
4-Bromo-3-chlorophenyl isothiocyanate
Formula: C₇H₃BrClNS
MDL Number: MFCD00041071
CAS Number: 32118-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR