4-Bromo-2-chlorophenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008746-1G 008746-5G
FLUO008746-1GEA 0 GBP
FLUO008746-1G FLUO008746-5G
4-Bromo-2-chlorophenyl isothiocyanate
4-Bromo-2-chlorophenyl isothiocyanate
Formula: C₇H₃BrClNS
Boiling Pt: 134 °C (0.5 mmHg)
Melting Pt: 46…49 °C
MDL Number: MFCD00041051
CAS Number: 98041-69-1
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR