N-Ethyl-4-(trifluoromethoxy)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008423-1G 008423-5G
FLUO008423-1GEA 0 GBP
FLUO008423-1G FLUO008423-5G
N-Ethyl-4-(trifluoromethoxy)aniline
N-Ethyl-4-(trifluoromethoxy)aniline
Formula: C₉H₁₀F₃NO
MDL Number: MFCD00203992
CAS Number: 175278-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR