2-Amino-6-fluorobenzothiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Fluorobenzothiazol-2-ylamine, 6-Fluorobenzo[d]thiazol-2-amine, 6-Fluoro-1,3-benzothiazol-2-amine

008164-10G 008164-50G 008164-1G 008164-5G
FLUO008164-10GEA 0 GBP
FLUO008164-10G FLUO008164-50G FLUO008164-1G FLUO008164-5G
2-Amino-6-fluorobenzothiazole
2-Amino-6-fluorobenzothiazole
Formula: C₇H₅FN₂S
MW: 168.19 g/mol
Boiling Pt: 304
Melting Pt: 183
Density: 1.59 g/cm³ (20 °C)
MDL Number: MFCD00013336
CAS Number: 348-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR