5-Fluoro-2-nitrobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008089-5G
008089-25G
FLUO008089-5GEA
0
GBP
FLUO008089-5G
FLUO008089-25G
5-Fluoro-2-nitrobenzoic acid
5-Fluoro-2-nitrobenzoic acid
Formula:
C₇H₄FNO₄ |
MDL Number:
MFCD00055635 CAS Number: 320-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...