Dipropyl sulphone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(Propylsulphonyl)propane

008053-5G 008053-25G
FLUO008053-5GEA 0 GBP
FLUO008053-5G FLUO008053-25G
Dipropyl sulphone
Dipropyl sulphone
Formula: C₆H₁₄O₂S
MW: 150.24 g/mol
Boiling Pt: 70…73 °C (0.3 mmHg)
Melting Pt: 29.5 °C
MDL Number: MFCD00007570
CAS Number: 598-03-8
EINECS: 209-913-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR