5-Amino-2,2-difluoro-1,3-benzodioxole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,2-Difluoro-1,3-benzodioxol-5-amine, 3,4-(Difluoromethylenedioxy)aniline

007844-5G 007844-10G
FLUO007844-5GEA 0 GBP
FLUO007844-5G FLUO007844-10G
5-Amino-2,2-difluoro-1,3-benzodioxole
5-Amino-2,2-difluoro-1,3-benzodioxole
Formula: C₇H₅F₂NO₂
Boiling Pt: 119 °C (35 mmHg)
MDL Number: MFCD00190144
CAS Number: 1544-85-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR