4-Chloro-3-(trifluoromethyl)benzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007806-5G 007806-10G 007806-1G
FLUO007806-5GEA 0 GBP
FLUO007806-5G FLUO007806-10G FLUO007806-1G
4-Chloro-3-(trifluoromethyl)benzaldehyde
4-Chloro-3-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
MDL Number: MFCD00040722
CAS Number: 34328-46-6
EINECS: 251-940-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR