2-Chloro-3-(trifluoromethyl)benzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007804-5G 007804-1G
FLUO007804-5GEA 0 GBP
FLUO007804-5G FLUO007804-1G
2-Chloro-3-(trifluoromethyl)benzaldehyde
2-Chloro-3-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
MDL Number: MFCD00792414
CAS Number: 93118-03-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR