Tris(pentafluorophenyl)phosphine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Perfluorotriphenylphosphine, Tris(perfluorophenyl)phosphine

007754-1G 007754-5G
FLUO007754-1GEA 0 GBP
FLUO007754-1G FLUO007754-5G
Tris(pentafluorophenyl)phosphine
Tris(pentafluorophenyl)phosphine
Formula: C₁₈F₁₅P
MDL Number: MFCD00079654
CAS Number: 1259-35-4
EINECS: 215-021-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR