Tris(4-fluorophenyl)phosphine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007752-1G 007752-5G
FLUO007752-1GEA 0 GBP
FLUO007752-1G FLUO007752-5G
Tris(4-fluorophenyl)phosphine
Tris(4-fluorophenyl)phosphine
Formula: C₁₈H₁₂F₃P
MDL Number: MFCD00013553
CAS Number: 18437-78-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR