(Trifluoromethyl)trimethylsilane
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
trimethyl(trifluoromethyl)silane
007685-25G
007685-10G
007685-100G
FLUO007685-25GEA
0
GBP
FLUO007685-25G
FLUO007685-10G
FLUO007685-100G
(Trifluoromethyl)trimethylsilane
(Trifluoromethyl)trimethylsilane
Formula:
C₄H₉F₃Si MW: 142.2 g/mol Boiling Pt: 55.5 °C (1013 hPa) Density: 0,95 g/cm³ (20 °C) |
MDL Number:
MFCD00145454 CAS Number: 81290-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...