1-(4-(Trifluoromethyl)phenyl)imidazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007650-1G 007650-5G
FLUO007650-1GEA 0 GBP
FLUO007650-1G FLUO007650-5G
1-(4-(Trifluoromethyl)phenyl)imidazole
1-(4-(Trifluoromethyl)phenyl)imidazole
Formula: C₁₀H₇F₃N₂
MDL Number: MFCD00060493
CAS Number: 25371-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR