1-(3-(Trifluoromethyl)phenyl)imidazole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
007649-1G
007649-5G
FLUO007649-1GEA
0
GBP
FLUO007649-1G
FLUO007649-5G
1-(3-(Trifluoromethyl)phenyl)imidazole
1-(3-(Trifluoromethyl)phenyl)imidazole
Formula:
C₁₀H₇F₃N₂ MW: 212.17 g/mol |
MDL Number:
MFCD00060487 CAS Number: 25371-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...