3-(Trifluoromethyl)benzyl mercaptan

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Trifluoromethyl)benzylthiol

007611-1G 007611-5G 007611-25G
FLUO007611-1GEA 0 GBP
FLUO007611-1G FLUO007611-5G FLUO007611-25G
3-(Trifluoromethyl)benzyl mercaptan
3-(Trifluoromethyl)benzyl mercaptan
Formula: C₈H₇F₃S
MW: 192.2 g/mol
Boiling Pt: 69…70 °C (1013 hPa)
MDL Number: MFCD00042435
CAS Number: 25697-55-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR