4-(Trifluoromethyl)benzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
007604-5G
007604-25G
007604-100G
FLUO007604-5GEA
0
GBP
FLUO007604-5G
FLUO007604-25G
FLUO007604-100G
4-(Trifluoromethyl)benzylamine
4-(Trifluoromethyl)benzylamine
Formula:
C₈H₈F₃N Boiling Pt: 95…97 °C (1013 hPa) Density: 1,229 g/cm³ (20 °C) |
MDL Number:
MFCD00010220 CAS Number: 3300-51-4 EINECS: 221-971-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...