3-(Trifluoromethyl)benzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
007603-5G
007603-25G
FLUO007603-5GEA
0
GBP
FLUO007603-5G
FLUO007603-25G
3-(Trifluoromethyl)benzylamine
3-(Trifluoromethyl)benzylamine
Formula:
C₈H₈F₃N MW: 175.15 g/mol Boiling Pt: 72…74 °C (13 mmHg) Density: 1.22 g/cm³ (20 °C) |
MDL Number:
MFCD00008117 CAS Number: 2740-83-2 EINECS: 220-367-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...