2-(Trifluoromethyl)benzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-(Trifluoromethyl)phenyl)methanamine

007602-25G 007602-5G
FLUO007602-25GEA 0 GBP
FLUO007602-25G FLUO007602-5G
2-(Trifluoromethyl)benzylamine
2-(Trifluoromethyl)benzylamine
Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 108 °C (64 mmHg)
MDL Number: MFCD00010297
CAS Number: 3048-01-9
EINECS: 221-258-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR