1,2,2,2-Tetrafluoro-1-iodoethane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007354-1G 007354-10G 007354-25G
FLUO007354-1GEA 0 GBP
FLUO007354-1G FLUO007354-10G FLUO007354-25G
1,2,2,2-Tetrafluoro-1-iodoethane
1,2,2,2-Tetrafluoro-1-iodoethane
Formula: C₂HF₄I
MW: 227.93 g/mol
MDL Number: MFCD00155949
CAS Number: 3831-49-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR