Silver hexafluoroarsenate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

007269-5G 007269-25G
FLUO007269-5GEA 0 GBP
FLUO007269-5G FLUO007269-25G
Silver hexafluoroarsenate
Silver hexafluoroarsenate
Formula: AgAsF₆
MDL Number: MFCD00014142
CAS Number: 12005-82-2
UN: 1557
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR