N-Methyl-4-(trifluoromethoxy)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006837-1G 006837-5G
FLUO006837-1GEA 0 GBP
FLUO006837-1G FLUO006837-5G
N-Methyl-4-(trifluoromethoxy)aniline
N-Methyl-4-(trifluoromethoxy)aniline
Formula: C₈H₈F₃NO
MDL Number: MFCD00052332
CAS Number: 41419-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR