2-Methoxy-5-(trifluoromethyl)pyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF660.1 006762-1G 006762-5G
CHMPCFF660.1EA 0 GBP
CHMPCFF660.1 FLUO006762-1G FLUO006762-5G
2-Methoxy-5-(trifluoromethyl)pyridine
2-Methoxy-5-(trifluoromethyl)pyridine
Formula: C₇H₆F₃NO
MDL Number: MFCD00153203
CAS Number: 175277-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR