4-Fluoro-3-(trifluoromethyl)benzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Fluoro-3-(trifluoromethyl)phenyl)methanamine

006421-1G 006421-5G 006421-10G
FLUO006421-1GEA 0 GBP
FLUO006421-1G FLUO006421-5G FLUO006421-10G
4-Fluoro-3-(trifluoromethyl)benzylamine
4-Fluoro-3-(trifluoromethyl)benzylamine
Formula: C₈H₇F₄N
MW: 193.14 g/mol
Boiling Pt: 101 °C (1013 hPa)
MDL Number: MFCD00070814
CAS Number: 67515-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR