4-Fluoro-2-(trifluoromethyl)benzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006385-5G 006385-25G
FLUO006385-5GEA 0 GBP
FLUO006385-5G FLUO006385-25G
4-Fluoro-2-(trifluoromethyl)benzaldehyde
4-Fluoro-2-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₄O
Boiling Pt: 114 °C (1013 hPa)
Density: 1.4 g/cm³ (20 °C)
MDL Number: MFCD00061313
CAS Number: 90176-80-0
UN: 1989
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR