2-Fluoro-6-(trifluoromethyl)benzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006383-1G 006383-5G 006383-10G
FLUO006383-1GEA 0 GBP
FLUO006383-1G FLUO006383-5G FLUO006383-10G
2-Fluoro-6-(trifluoromethyl)benzaldehyde
2-Fluoro-6-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₄O
MDL Number: MFCD00061273
CAS Number: 60611-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR