4-Fluorophthalic anhydride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006356-5G
006356-25G
FLUO006356-5GEA
0
GBP
FLUO006356-5G
FLUO006356-25G
4-Fluorophthalic anhydride
4-Fluorophthalic anhydride
Formula:
C₈H₃FO₃ Boiling Pt: 260 °C (1013 hPa) Melting Pt: 79…80 °C Density: 1,5 g/cm³ (20 °C) |
MDL Number:
MFCD00191363 CAS Number: 319-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...