1-(3-Fluorophenyl)-2-thiourea

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: N-(3-Fluorophenyl)thiourea, 3-Fluorophenylthiourea

006348-5G 006348-25G
FLUO006348-5GEA 0 GBP
FLUO006348-5G FLUO006348-25G
1-(3-Fluorophenyl)-2-thiourea
1-(3-Fluorophenyl)-2-thiourea
Formula: (C₆H₄F)NC(S)NH₂
MW: 170.21 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 116 °C
MDL Number: MFCD00041167
CAS Number: 458-05-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR