2,4-Bis(trifluoromethyl)benzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006011-1G
006011-5G
FLUO006011-1GEA
0
GBP
FLUO006011-1G
FLUO006011-5G
2,4-Bis(trifluoromethyl)benzonitrile
2,4-Bis(trifluoromethyl)benzonitrile
Formula:
C₉H₃F₆N MW: 239.12 g/mol |
MDL Number:
MFCD00061225 CAS Number: 177952-38-4 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...