4-Bromo-2-nitroaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005887-1G
005887-100G
005887-25G
FLUO005887-1GEA
0
GBP
FLUO005887-1G
FLUO005887-100G
FLUO005887-25G
4-Bromo-2-nitroaniline
4-Bromo-2-nitroaniline
Formula:
C₆H₅BrN₂O₂ Boiling Pt: 309 °C (1013 hPa) Melting Pt: 110…113 °C Density: 1,81 g/cm³ (20 °C) |
MDL Number:
MFCD00041312 CAS Number: 875-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...