3-(Trifluoromethyl)-p-anisidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005826-5G 005826-10G 005826-1G
FLUO005826-5GEA 0 GBP
FLUO005826-5G FLUO005826-10G FLUO005826-1G
3-(Trifluoromethyl)-p-anisidine
3-(Trifluoromethyl)-p-anisidine
Formula: C₈H₈F₃NO
Boiling Pt: 112 °C (18 mmHg)
Melting Pt: 59…60 °C
MDL Number: MFCD00043460
CAS Number: 393-15-7
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR