2,3-Difluoro-4-methylbenzyl bromide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005758-1G 005758-5G
FLUO005758-1GEA 0 GBP
FLUO005758-1G FLUO005758-5G
2,3-Difluoro-4-methylbenzyl bromide
2,3-Difluoro-4-methylbenzyl bromide
Formula: C₈H₇BrF₂
MDL Number: MFCD01631647
CAS Number: 261763-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR