5-Chloro-2-fluorobenzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005629-1G
005629-5G
FLUO005629-1GEA
0
GBP
FLUO005629-1G
FLUO005629-5G
5-Chloro-2-fluorobenzylamine
5-Chloro-2-fluorobenzylamine
Formula:
C₇H₇ClFN Boiling Pt: 211 °C (1013 hPa) Melting Pt: 60 °C |
MDL Number:
MFCD01631440 CAS Number: 261723-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...