2-Chloro-3,6-difluorobenzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005598-5G
005598.10G
005598-1G
FLUO005598-5GEA
0
GBP
FLUO005598-5G
FLUO005598.10G
FLUO005598-1G
2-Chloro-3,6-difluorobenzylamine
2-Chloro-3,6-difluorobenzylamine
Formula:
C₇H₆ClF₂N Boiling Pt: 207 °C (1013 hPa) Melting Pt: 73 °C |
MDL Number:
MFCD01631449 CAS Number: 261762-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...