Dibenzothiophene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Dibenzo[b,d]thiophene
005547-25G
005547-100G
FLUO005547-25GEA
0
GBP
FLUO005547-25G
FLUO005547-100G
Dibenzothiophene
Dibenzothiophene
Formula:
C₁₂H₈S Boiling Pt: 332 °C (1013 hPa) Melting Pt: 95…98 °C Density: 1,1295 g/cm³ (125 °C) Flash Pt: 170 °C |
MDL Number:
MFCD00004969 CAS Number: 132-65-0 EINECS: 205-072-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...