3,4-Dichlorobenzoylacetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(3,4-Dichlorophenyl)-3-oxopropanenitrile

005493-1G 005493-5G
FLUO005493-1GEA 0 GBP
FLUO005493-1G FLUO005493-5G
3,4-Dichlorobenzoylacetonitrile
3,4-Dichlorobenzoylacetonitrile
Formula: C₉H₅Cl₂NO
MDL Number: MFCD00067919
CAS Number: 4640-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR