1-(4-Fluorophenyl)-2-thiourea

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: N-(4-Fluorophenyl)thiourea, 4-Fluorophenylthiourea

005417-5G 005417-25G 005417-100G
FLUO005417-5GEA 0 GBP
FLUO005417-5G FLUO005417-25G FLUO005417-100G
1-(4-Fluorophenyl)-2-thiourea
1-(4-Fluorophenyl)-2-thiourea
Formula: (C₆H₄F)NHC(S)NH₂
MW: 170.21 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 169…170 °C
MDL Number: MFCD00041180
CAS Number: 459-05-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR