N-Benzylmaleimide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005320-5G
005320-25G
FLUO005320-5GEA
0
GBP
FLUO005320-5G
FLUO005320-25G
N-Benzylmaleimide
N-Benzylmaleimide
Formula:
C₁₁H₉NO₂ Boiling Pt: 339 °C (1013 hPa) Melting Pt: 70…72 °C Density: 1,296 g/cm³ (20 °C) |
MDL Number:
MFCD00014540 CAS Number: 1631-26-1 EINECS: 216-631-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...