2-Chloro-4-(trifluoromethyl)pyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF600.0.5
005218-1G
005218-5G
CHMPCFF600.0.5EA
0
GBP
CHMPCFF600.0.5
FLUO005218-1G
FLUO005218-5G
2-Chloro-4-(trifluoromethyl)pyridine
2-Chloro-4-(trifluoromethyl)pyridine
Formula:
C₆H₃ClF₃N Boiling Pt: 146…147 °C (1013 hPa) Melting Pt: –1 °C Density: 1.411 g/cm³ (20 °C) |
MDL Number:
MFCD00042224 CAS Number: 81565-18-6 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...