2,4-Bis(trifluoromethyl)benzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004998-1G 004998-5G
FLUO004998-1GEA 0 GBP
FLUO004998-1G FLUO004998-5G
2,4-Bis(trifluoromethyl)benzaldehyde
2,4-Bis(trifluoromethyl)benzaldehyde
Formula: C₉H₄F₆O
MDL Number: MFCD00083535
CAS Number: 59664-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR