3,5-Dichloro-4-iodobenzotrifluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004721-1G
004721-5G
FLUO004721-1GEA
0
GBP
FLUO004721-1G
FLUO004721-5G
3,5-Dichloro-4-iodobenzotrifluoride
3,5-Dichloro-4-iodobenzotrifluoride
Formula:
C₇H₂Cl₂F₃I MW: 340.9 g/mol |
MDL Number:
MFCD00084997 CAS Number: 175205-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...