4-Fluoro-o-phenylenediamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,4-Diamino-1-fluorobenzene, 4-Fluorobenzene-1,2-diamine, 1,2-Diamino-4-fluorobenzene, 4-Fluoro-1,2-phenylenediamine
004630-25G
004630-5G
FLUO004630-25GEA
0
GBP
FLUO004630-25G
FLUO004630-5G
4-Fluoro-o-phenylenediamine
4-Fluoro-o-phenylenediamine
Formula:
C₆H₇FN₂ MW: 126.13 g/mol Boiling Pt: 250 Melting Pt: 92 |
MDL Number:
MFCD00042228 CAS Number: 367-31-7 EINECS: 206-691-7 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...