2-Chloro-5-(trifluoromethyl)pyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF601.1 004552-100G 004552-25G
CHMPCFF601.1EA 0 GBP
CHMPCFF601.1 FLUO004552-100G FLUO004552-25G
2-Chloro-5-(trifluoromethyl)pyridine
2-Chloro-5-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃N
Boiling Pt: 152 °C (1013 hPa)
Melting Pt: 30…31 °C
Density: 1.41 g/cm³ (20 °C)
MDL Number: MFCD00042225
CAS Number: 52334-81-3
EINECS: 257-856-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR