5-Chloro-2-iodobenzotrifluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004485-25G
004485-100G
FLUO004485-25GEA
0
GBP
FLUO004485-25G
FLUO004485-100G
5-Chloro-2-iodobenzotrifluoride
5-Chloro-2-iodobenzotrifluoride
Formula:
C₇H₃ClF₃I Boiling Pt: 232 °C (1013 hPa) Melting Pt: 29…31 °C Density: 1.97 g/cm³ (20 °C) |
MDL Number:
MFCD00042579 CAS Number: 23399-77-1 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...