3-Chloro-4-fluorobenzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004443-5G 004443-25G
FLUO004443-5GEA 0 GBP
FLUO004443-5G FLUO004443-25G
3-Chloro-4-fluorobenzylamine
3-Chloro-4-fluorobenzylamine
Formula: C₇H₇ClFN
MW: 159.59 g/mol
Boiling Pt: 104…105 °C (15 mmHg)
MDL Number: MFCD00042507
CAS Number: 72235-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR