2-Bromo-5-(trifluoromethyl)pyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004308-1G
004308-5G
004308-25G
FLUO004308-1GEA
0
GBP
FLUO004308-1G
FLUO004308-5G
FLUO004308-25G
2-Bromo-5-(trifluoromethyl)pyridine
2-Bromo-5-(trifluoromethyl)pyridine
Formula:
C₆H₃BrF₃N MW: 226 g/mol |
MDL Number:
MFCD00153086 CAS Number: 50488-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...