4-Bromo-3,5-dichlorobenzotrifluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004178-1G
004178-5G
004178-25G
FLUO004178-1GEA
0
GBP
FLUO004178-1G
FLUO004178-5G
FLUO004178-25G
4-Bromo-3,5-dichlorobenzotrifluoride
4-Bromo-3,5-dichlorobenzotrifluoride
Formula:
C₇H₂BrCl₂F₃ MW: 293.9 g/mol |
MDL Number:
MFCD00221020 CAS Number: 118754-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...