3-Amino-4-(trifluoromethyl)pyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004035-500MG 004035-1G 004035-2.5G
FLUO004035-500MGEA 0 GBP
FLUO004035-500MG FLUO004035-1G FLUO004035-2.5G
3-Amino-4-(trifluoromethyl)pyridine
3-Amino-4-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MDL Number: MFCD00173960
CAS Number: 175204-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR